Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DUX4 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $526.00
Specifications
Antigen | DUX4 |
---|---|
Clone | 2142A |
Dilution | Western Blot 0.1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 3-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9535SP
![]() |
R&D Systems
MAB9535SP |
25 μg |
Each for $214.00
|
|
|||||
MAB9535100
![]() |
R&D Systems
MAB9535100 |
100 μg |
Each for $526.00
|
|
|||||
Description
DUX4 Monoclonal specifically detects DUX4 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
DUX4 | |
Western Blot 0.1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 3-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
100288687 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
2142A | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
double homeobox 4, double homeobox protein 10, double homeobox protein 4, double homeobox protein 4/10, double homeobox protein DUX10, DUX10, DUX4L1 | |
Human DUX4 synthetic peptide | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human DUX4 in direct ELISAs and Western blots. |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title