Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DUX4/DUX4c Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB95351SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB95351SP 25 μg
MAB95351100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB95351SP Supplier R&D Systems Supplier No. MAB95351SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody has been used in 2 publications

DUX4/DUX4c Monoclonal specifically detects DUX4/DUX4c in Human samples. It is validated for Western Blot.

Specifications

Antigen DUX4/DUX4c
Applications Western Blot
Classification Monoclonal
Clone 2143C
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q9UBX2
Gene Alias DUX10, DUX4, DUX4L, DUX4L1, DUX4L9
Host Species Rabbit
Immunogen Human DUX4 synthetic peptide containing sequences within amino acids 200-300 of the human DUX4 protein Accession # Q9UBX2
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Test Specificity Detects human DUX4/DUX4c in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.