Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Dynactin Subunit 1/DCTN1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6657
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6657 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6657 Supplier R&D Systems Supplier No. AF6657
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Dynactin Subunit 1/DCTN1 Polyclonal specifically detects Dynactin Subunit 1/DCTN1 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Dynactin Subunit 1/DCTN1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q14203
Gene Alias DAP-150, DCTN1, DP-150, Dynactin 1, dynactin 1 (p150, Glued (Drosophila) homolog), dynactin subunit 1, glued homolog, Drosophila), HMN7B, p135, p150 Glued (Drosophila) homolog, p150-glued
Gene Symbols DCTN1
Host Species Sheep
Immunogen E. coli-derived recombinant human Dynactin Subunit 1/DCTN1 Ala1145-Ser1278 Accession # Q14203
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1639
Test Specificity Detects human Dynactin Subunit 1/DCTN1 in direct ELISAs and human, mouse and rat Dynactin Subunit 1/DCTN1 in Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.