Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Dynactin Subunit 2/DCTN2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6850SP
This item is not returnable.
View return policy
Description
Dynactin Subunit 2/DCTN2/DCTN-50 Monoclonal specifically detects Dynactin Subunit 2/DCTN2/DCTN-50 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Dynactin Subunit 2/DCTN2/DCTN-50 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Q13561 | |
DCTN2 | |
E. coli-derived recombinant human Dynactin Subunit 2/DCTN2 Asp277-Lys401 Accession # Q13561 | |
25 μg | |
Primary | |
Detects human Dynactin Subunit 2/DCTN2 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Dynactin Subunit 1/DCTN1 is observed. | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
696206 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DCTN2, DCTN-50, DCTN5050 kD dynein-associated polypeptide, dynactin 2 (p50), dynactin complex 50 kD subunit, Dynactin complex 50 kDa subunit, dynactin subunit 2,50 kDa dynein-associated polypeptide, DYNAMITIN, p50 dynamitin, RBP50 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10540 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction