Learn More
Human E-Cadherin Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | E-Cadherin |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 180224 |
| Conjugate | Alexa Fluor 488 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gene Accession No. | P12830 |
| Gene Alias | Arc-1, CAD1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDH1, CDHE, cell-CAM 120/80, Cell-CAM120/80, ECAD, E-cadherin, ECadherin, Epithelial cadherin, LCAM, L-CAM, UVOE-Cadherin, uvomorulin |
| Gene Symbols | CDH1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.