Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human E-Cadherin Protein

Catalog No. 8505ec050
Click to view available options
Quantity:
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human E-Cadherin Protein is derived from HEK293. The Recombinant Human E-Cadherin Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P12830
For Use With (Application) Bioactivity
Gene ID (Entrez) 999
Molecular Weight (g/mol) Observed M.W.: 74-92kDa, reducing conditions, Theoretical M.W.: 61kDa
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human E-Cadherin protein Asp155-Ile707,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Arc-1, CAD1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDH1, CDHE, cell-CAM 120/80, Cell-CAM120/80, ECAD, E-cadherin, ECadherin, Epithelial cadherin, LCAM, L-CAM, UVOE-Cadherin, uvomorulin
Common Name E-Cadherin
Gene Symbol CDH1
Biological Activity Measured by the ability of the immobilized protein to support the adhesion of the MCF-7 human breast cancer cells. The ED50 for this effect is typically 0.2-1μg/mL
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Asp155-Ile707, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.