Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human E-Selectin/CD62E Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BBA26200
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BBA26200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BBA26200 Supplier R&D Systems Supplier No. BBA26200
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 7 publications

E-Selectin/CD62E Monoclonal specifically detects E-Selectin/CD62E in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Adhesion Blockade, Immunofluorescence, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen E-Selectin/CD62E
Applications Flow Cytometry, Immunoprecipitation, Adhesion Blockade, Immunofluorescence, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone BBIG-E1
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, Immunoprecipitation, Adhesion Blockade, Immunofluorescence, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative
Gene Alias CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM1, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, SELE, selectin E
Gene Symbols SELE
Host Species Mouse
Immunogen Activated HUVEC human umbilical vein endothelial cells
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6401
Test Specificity This antibody binds to CHO cells transfected with human E-Selectin. It does not bind to COS cells transfected with human PECAM-1, P-Selectin, L-Selectin, VCAM-1, or ICAM-1.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.