Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human E-Selectin/CD62E Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF724
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF724 100 μg
AF724SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF724 Supplier R&D Systems Supplier No. AF724
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

E-Selectin/CD62E Polyclonal specifically detects E-Selectin/CD62E in Human samples. It is validated for Western Blot, Adhesion Blockade.

Specifications

Antigen E-Selectin/CD62E
Applications Western Blot, Adhesion Blockade
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Adhesion Blockade
Gene Alias CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM1, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, SELE, selectin E
Gene Symbols SELE
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human E-Selectin
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6401
Test Specificity Selected for its ability to inhibit the block U937 cell adhesion to immobilized rhE-Selectin. In direct ELISAs, less than 5% cross-reactivity with rmE-Selectin and rrE-Selectin is observed and less than 1% cross-reactivity with rhP-Selectin and rhL-Selectin is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.