Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human E-Selectin/P-Selectin (CD62E/P) PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6169P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6169P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6169P Supplier R&D Systems Supplier No. FAB6169P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

E-Selectin (CD62E)/P-Selectin (CD62P) Monoclonal specifically detects E-Selectin (CD62E)/P-Selectin (CD62P) in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen E-Selectin (CD62E)/P-Selectin (CD62P)
Applications Flow Cytometry
Classification Monoclonal
Clone BBIG-E6 (13D5)
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias CD62E, CD62P, ELAM, ELAM1, ESEL, Eselectin, GMP140, GRMP, LECAM2, PADGEM, PSEL, PSelectin, SELE, SELP
Host Species Mouse
Immunogen Activated HUVEC human umbilical vein endothelial cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6401
Test Specificity Detects human E-Selectin/P-Selectin (CD62E/P). Binds to COS cells transfected with human E-Selectin or human P-Selectin. It does not bind to CHO cells transfected with human ICAM-1, L-Selectin, PECAM-1 or VCAM-1.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.