Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human E2F-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5119SP
This item is not returnable.
View return policy
Description
E2F-2 Polyclonal specifically detects E2F-2 in Human samples. It is validated for Western Blot.Specifications
E2F-2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
E2F transcription factor 2, E2F2, E2F-2transcription factor E2F2 | |
Goat | |
Affinity Purified | |
RUO | |
1870 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q14209 | |
E2F2 | |
E. coli-derived recombinant human E2F-2 Glu308-Asn437 Accession # Q14209 | |
25 μg | |
Primary | |
Detects human E2F-2 in Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction