Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human E2F-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5119
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5119 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5119 Supplier R&D Systems Supplier No. AF5119
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

E2F-2 Polyclonal specifically detects E2F-2 in Human samples. It is validated for Western Blot.

Specifications

Antigen E2F-2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q14209
Gene Alias E2F transcription factor 2, E2F2, E2F-2transcription factor E2F2
Gene Symbols E2F2
Host Species Goat
Immunogen E. coli-derived recombinant human E2F-2 Glu308-Asn437 Accession # Q14209
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1870
Test Specificity Detects human E2F-2 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.