Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human E2F-4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5139
This item is not returnable.
View return policy
Description
E2F-4 Polyclonal specifically detects E2F-4 in Human samples. It is validated for Western Blot.Specifications
E2F-4 | |
Polyclonal | |
Unconjugated | |
Q16254 | |
E2F4 | |
E. coli-derived recombinant human E2F-4 Lys178-Asp306 Accession # Q16254 | |
100 μg | |
Primary | |
Detects endogenous human E2F-4 in Western bots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
E2F transcription factor 4, p107/p130-binding, E2F4, E2F-4p107/p130-binding protein, transcription factor E2F4 | |
Goat | |
Affinity Purified | |
RUO | |
1874 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction