Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human E6AP/UBE3A Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
$205.85 - $479.23
Specifications
Antigen | E6AP/UBE3A |
---|---|
Clone | 1077332 |
Dilution | Western Blot 2 μg/mL, Simple Western 20 μg/mL |
Applications | Western Blot, Simple Western |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11498100
![]() |
Novus Biologicals™
MAB11498100 |
100 μg |
Each for $479.23
|
|
|||||
MAB11498SP
![]() |
Novus Biologicals™
MAB11498SP |
25 μg |
Each for $205.85
|
|
|||||
Description
E6AP/UBE3A Monoclonal antibody specifically detects E6AP/UBE3A in Human samples. It is validated for Western Blot,Simple WesternSpecifications
E6AP/UBE3A | |
Western Blot 2 μg/mL, Simple Western 20 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
ANCR, ANCREPVE6AP, AS, CTCL tumor antigen se37-2, E6AP, E6-AP, E6AP ubiquitin-protein ligase, EC 6.3.2.-, EPVE6AP, FLJ26981, HPVE6A, human papilloma virus E6-associated protein, Human papillomavirus E6-associated protein, Oncogenic protein-associated protein E6-AP, Renal carcinoma antigen NY-REN-54, UBE3A, ubiquitin protein ligase E3A, ubiquitin-protein ligase E3A | |
Spodoptera frugiperda, Sf 21 (baculovirus)-derived human E6AP/UBE3A, Met1-Leu875, Accession # Q05086 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
1077332 | |
Western Blot, Simple Western | |
Unconjugated | |
Mouse | |
Human | |
Q05086 | |
7337 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze/thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title