Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human EAAT1/GLAST-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6048100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6048100 100 μg
MAB6048SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB6048100 Supplier R&D Systems Supplier No. MAB6048100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

EAAT1/GLAST-1/SLC1A3 Monoclonal specifically detects EAAT1/GLAST-1/SLC1A3 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen EAAT1/GLAST-1/SLC1A3
Applications Immunohistochemistry
Classification Monoclonal
Clone 482420
Conjugate Unconjugated
Dilution Immunohistochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Gene Accession No. P43003
Gene Alias EA6, EA6FLJ25094, EAAT1GLAST-1, excitatory amino acid transporter 1, GLAST1, GLAST-1, GLASTGLAST1, SLC1A3, Sodium-dependent glutamate/aspartate transporter 1, solute carrier family 1 (glial high affinity glutamate transporter), member 3, Solute carrier family 1 member 3
Host Species Mouse
Immunogen E. coli-derived recombinant human EAAT1/GLAST-1, His146-Ser237, Accession # P43003
Purification Method Protein A or G purified from ascites
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6507
Test Specificity Detects human EAAT1/GLAST-1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.