Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EBI2 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB102721T100UG
This item is not returnable.
View return policy
Description
EBI2/GPR183 Monoclonal specifically detects EBI2/GPR183 in Human samples. It is validated for Flow Cytometry.Specifications
EBI2/GPR183 | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
EBI2Epstein-Barr virus induced gene 2 (lymphocyte-specific G protein-coupledreceptor), EBV-induced G-protein coupled receptor 2, Epstein-Barr virus-induced G-protein coupled receptor 2, G protein-coupled receptor 183, GPR183, G-protein coupled receptor 183 | |
Mouse myeloma cell line NS0-derived transfected with recombinant human EBI2, Accession # P32249 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
486617 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P32249 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1880 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction