Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ECE-1 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems IC17841U100UG
This item is not returnable.
View return policy
Description
ECE-1 Monoclonal specifically detects ECE-1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
ECE-1 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P42892 | |
ECE1 | |
Mouse myeloma cell line NS0-derived recombinant human ECE-1 Gln90-Trp770 Accession # P42892 | |
RUO | |
1889 | |
Human | |
IgG1 |
Flow Cytometry | |
303913 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
EC 3.4.24, EC 3.4.24.71, ECE, ECE1, ECE-1, endothelin converting enzyme 1, endothelin-converting enzyme 1 | |
Rat | |
100 μg | |
Primary | |
Detects human ECE-1 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction