Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ECM-1 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$200.50 - $549.50

Specifications

Antigen ECM-1/Secretory Component P85
Clone 966231
Dilution Western Blot 2 ug/mL, ELISA
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB39371SP
GREENER_CHOICE
View Documents
R&D Systems
MAB39371SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB39371100
GREENER_CHOICE
View Documents
R&D Systems
MAB39371100
100 μg
Each for $549.50
Only null left
Add to Cart
 
Description

Description

ECM-1/Secretory Component P85 Monoclonal specifically detects ECM-1/Secretory Component P85 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

ECM-1/Secretory Component P85
Western Blot 2 ug/mL, ELISA
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
ECM1, extracellular matrix protein 1, p85, Secretory Component Glycoprotein, Secretory Component P85
Mouse myeloma cell line NS0-derived recombinant human ECM-1 Ala20-Glu540 Accession # Q16610
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human ECM-1 in direct ELISAs and Western blots.
966231
Western Blot, ELISA
Unconjugated
Mouse
Human
Q16610
1893
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

RUO

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.