Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ECM1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF3937
This item is not returnable.
View return policy
Description
ECM-1/Secretory Component P85 Polyclonal specifically detects ECM-1/Secretory Component P85 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
ECM-1/Secretory Component P85 | |
Polyclonal | |
Unconjugated | |
AAH23505 | |
ECM1 | |
Mouse myeloma cell line NS0-derived recombinant human ECM-1 Ala20-Glu540 Accession # AAH23505 | |
100 μg | |
Primary | |
Detects human ECM-1 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse ECM-1 is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL | |
ECM1, extracellular matrix protein 1, p85, Secretory Component Glycoprotein, Secretory Component P85 | |
Sheep | |
Affinity Purified | |
RUO | |
1893 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction