Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ECM1 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF3937
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF3937 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF3937 Supplier R&D Systems Supplier No. BAF3937
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

ECM-1/Secretory Component P85 Polyclonal specifically detects ECM-1/Secretory Component P85 in Human samples. It is validated for Western Blot.

Specifications

Antigen ECM-1/Secretory Component P85
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. AAH23505
Gene Alias ECM1, extracellular matrix protein 1, p85, Secretory Component Glycoprotein, Secretory Component P85
Gene Symbols ECM1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human ECM-1 Ala20-Glu540 Accession # AAH23505
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1893
Test Specificity Detects human ECM-1 in Western blots. In Western blots, less than 10% cross-reactivity with recombinant mouse ECM-1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.