Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human EDA-A2/Ectodysplasin A2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF922
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF922 100 μg
AF922SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF922 Supplier R&D Systems Supplier No. AF922
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

EDA-A2/Ectodysplasin A2 Polyclonal specifically detects EDA-A2/Ectodysplasin A2 in Human samples. It is validated for Blockade of Receptor-ligand Interaction.

Specifications

Antigen EDA-A2/Ectodysplasin A2
Applications Blockade of Receptor-ligand Interaction
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Blockade of Receptor-ligand Interaction
Gene Accession No. NP_001005609
Gene Symbols EDA
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human EDA-A2/Ectodysplasin A2 Ala179-Ser389 Accession # NP_001005609
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1896
Test Specificity Detects human EDA-A2/Ectodysplasin A2 in direct ELISAs and Western blots. In Western blots, less than 10% cross-reactivity with recombinant human (rh) APRIL, rhBAFF, rhLIGHT, rhOX40 Ligand, and rhTRAIL is observed and less than 2% cross-reactivity with recombinant mouse EDA, rhTNF-alpha , rhVEGI, rhGITR Ligand, and rhFasLigand is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.