Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human EDA/Ectodysplasin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9222SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9222SP 25 μg
MAB9222 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9222SP Supplier R&D Systems Supplier No. MAB9222SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

EDA/Ectodysplasin Monoclonal specifically detects EDA/Ectodysplasin in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).

Specifications

Antigen EDA/Ectodysplasin
Applications ELISA
Classification Monoclonal
Clone 174806
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_001005609
Gene Alias ectodysplasin A, ED1, EDAA1, EDAA2, Tabby, XHED, XLHED
Gene Symbols EDA
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human EDA-A2/Ectodysplasin A2 Ala179-Ser389 Accession # NP_001005609
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1896
Test Specificity Detects human EDA/Ectodysplasin (EDA-A1 and EDA-A2) in ELISAs. In sandwich immunoassays, approximately 50% cross-reactivity with recombinant mouse EDA is observed, and no cross-reactivity with recombinant human (rh) TNF-alpha , rhXEDAR, or rhEDAR is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.