Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EDIL3 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6046SP
This item is not returnable.
View return policy
Description
EDIL3/DEL1 Polyclonal specifically detects EDIL3/DEL1 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
EDIL3/DEL1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DEL1, DEL1developmental endothelial locus-1, Developmentally-regulated endothelial cell locus 1 protein, EGF-like repeat and discoidin I-like domain-containing protein 3, EGF-like repeats and discoidin I-like domains 3, Integrin-binding protein DEL1, MGC26287 | |
Sheep | |
Affinity Purified | |
RUO | |
10085 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
O43854 | |
EDIL3 | |
Chinese hamster ovary cell line CHO-derived recombinant human EDIL3 Val17-Glu480 Accession # O43854 | |
25 μg | |
Primary | |
Detects human EDIL3 in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction