Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human EDNRA Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6538SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6538SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6538SP Supplier R&D Systems Supplier No. MAB6538SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

EDNRA/Endothelin R Type A Monoclonal specifically detects EDNRA/Endothelin R Type A in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen EDNRA/Endothelin R Type A
Applications Immunohistochemistry
Classification Monoclonal
Clone 485711
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_001948
Gene Alias EDNRA, endothelin receptor type A, endothelin-1 receptor, endothelin-1-specific receptor, ET1-specific type, ETAR, ETRA, G protein-coupled receptor, hET-AR
Gene Symbols EDNRA
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human EDRNA Accession # NP_001948
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1909
Test Specificity Detects human EDRNA in direct ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.