Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EDNRA Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6538
This item is not returnable.
View return policy
Description
EDNRA/Endothelin R Type A Monoclonal specifically detects EDNRA/Endothelin R Type A in Human samples. It is validated for Immunohistochemistry.Specifications
EDNRA/Endothelin R Type A | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
NP_001948 | |
EDNRA | |
NS0 mouse myeloma cell line transfected with human EDRNA Accession # NP_001948 | |
100 μg | |
Primary | |
Detects human EDRNA in direct ELISAs. | |
Human | |
Purified |
Immunohistochemistry | |
485711 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EDNRA, endothelin receptor type A, endothelin-1 receptor, endothelin-1-specific receptor, ET1-specific type, ETAR, ETRA, G protein-coupled receptor, hET-AR | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1909 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction