Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EG-VEGF/PK1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB12091SP
This item is not returnable.
View return policy
Description
EG-VEGF/PK1 Monoclonal specifically detects EG-VEGF/PK1 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| EG-VEGF/PK1 | |
| Monoclonal | |
| LYOPH | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P58294 | |
| PROK1 | |
| Mouse myeloma cell line NS0-derived recombinant human EG-VEGF/PK1 Ala20-Phe105 Accession # P58294 | |
| 25 μg | |
| Primary | |
| Detects human EG-VEGF/PK1 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| ELISA | |
| 188608 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EGVEGF, EG-VEGF, EGVEGFblack mamba toxin-related protein, Endocrine-gland-derived vascular endothelial growth factor, Mambakine, PK1mambakine, PRK1prokineticin-1, PROK1, prokineticin 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 84432 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction