Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human EGF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AB236NA
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB236NA 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AB236NA Supplier R&D Systems Supplier No. AB236NA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

EGF Polyclonal specifically detects EGF in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen EGF
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 1-3 ug/mL
Gene Accession No. P01133
Gene Alias beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), hEGF, HOMG4, pro-epidermal growth factor, URG, Urogastrone
Gene Symbols EGF
Host Species Goat
Immunogen E. coli-derived recombinant human EGF Asn971-Arg1023 Accession # P01133
Purification Method Protein A or G purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1950
Test Specificity Detects human EGF in direct ELISAs and Western blots. In direct ELISAs, less than 15% cross-reactivity with recombinant rat EGF and recombinant mouse EGF is observed. Does not neutralize the biological activity of recombinant human TGF-alpha .
Reconstitution Reconstitute at 1 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.