Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EGF Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 7 publications
Supplier: R&D Systems AF236SP
This item is not returnable.
View return policy
Description
EGF Polyclonal specifically detects EGF in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
EGF | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EGF | |
E. coli-derived recombinant human EGF | |
25 μg | |
Primary | |
Detects human EGF in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant rat EGF and 5% cross-reactivity with recombinant mouse EGF is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.040 - 0.480 ug/mL | |
beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), hEGF, HOMG4, pro-epidermal growth factor, URG, Urogastrone | |
Goat | |
Affinity Purified | |
RUO | |
1950 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction