Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EGFR Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB10951G
This item is not returnable.
View return policy
Description
EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Flow Cytometry.Specifications
EGFR | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, EGF R, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ErbB, ErbB1, ERBB1PIG61, HER1, HER-1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1956 | |
Human | |
Purified |
Flow Cytometry | |
423103 | |
Flow Cytometry 5 uL/10^6 cells | |
CAA25240 | |
EGFR | |
Mouse myeloma cell line NS0-derived recombinant human EGF R/ErbB1 Leu25-Ser645 Accession # CAA25240 | |
100 Tests | |
Primary | |
Detects human EGF R/ErbB1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) ErbB2, rhErbB3, or rhErbB4 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction