Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EGFR F(ab')2 (Research Grade Cetuximab Biosimilar) Antibody, Novus Biologicals™

Human Monoclonal Antibody
Supplier: R&D Systems MAB9577FAB2100
This item is not returnable.
View return policy
Description
EGFR Monoclonal antibody specifically detects EGFR in Human samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
EGFR | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
Human | |
Protein A or G purified from cell culture supernatant | |
RUO | |
1956 | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry | |
Hu1F2 | |
Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL | |
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, EGF R, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ErbB, ErbB1, ERBB1PIG61, HER1, HER-1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
Human EGFR/ErbB1 | |
100 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction