Learn More
Human EGFR (Research Grade Cetuximab Biosimilar) Biotinylated Antibody, R&D Systems™
Human Monoclonal Antibody
Supplier: R&D Systems FAB9577B100
This item is not returnable.
View return policy
Description
EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
EGFR | |
Monoclonal | |
Biotin | |
Supplied in a saline solution containing BSA and Sodium Azide. | |
Human | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human EGFR based on Cetuximab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Cetuximab. This product is for research use only. | |
Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Flow Cytometry, Immunocytochemistry | |
Hu1 | |
Flow Cytometry 10 uL/10^6 cells, Immunocytochemistry 8-25 ug/mL | |
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, EGF R, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ErbB, ErbB1, ERBB1PIG61, HER1, HER-1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
Human EGFR | |
100 Tests | |
1956 | |
Human | |
Supernatant |