Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human EGR1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB28181100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB28181100 100 μg
MAB28181SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB28181100 Supplier R&D Systems Supplier No. MAB28181100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

EGR1 Monoclonal specifically detects EGR1 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen EGR1
Applications Immunohistochemistry
Classification Monoclonal
Clone 406910
Conjugate Unconjugated
Dilution Immunohistochemistry 5-25 μ/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P18146
Gene Alias AT225Transcription factor ETR103, early growth response 1, early growth response protein 1, EGR-1, G0S30, KROX24, KROX-24, Nerve growth factor-induced protein A, NGFIA, NGFI-ATranscription factor Zif268, TIS8, ZIF-268, Zinc finger protein Krox-24, ZNF225, ZNF225Zinc finger protein 225
Host Species Rat
Immunogen E. coli-derived recombinant human EGR1, Gln282-Ser433, Accession No. P18146
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1958
Test Specificity Detects human EGR1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.