Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human eIF2 alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3997
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3997 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3997 Supplier R&D Systems Supplier No. AF3997
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

eIF2 alpha/EIF2S1 Polyclonal specifically detects eIF2 alpha/EIF2S1 in Human samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen eIF2 alpha/EIF2S1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL
Gene Accession No. Q53XC0
Gene Alias EIF2, EIF-2, EIF2A, EIF-2A, EIF-2alpha, EIF2S1, EIF2S1 eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa
Gene Symbols EIF2S1
Host Species Goat
Immunogen E. coli-derived recombinant human eIF2 alpha Met1-Asp315 Accession # Q53XC0
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1965
Test Specificity Detects human, mouse, and rat eIF2 alpha in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.