Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ELF3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB57871SP
This item is not returnable.
View return policy
Description
ELF3/ESE-1 Monoclonal specifically detects ELF3/ESE-1 in Human samples. It is validated for Immunohistochemistry.Specifications
ELF3/ESE-1 | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
P78245 | |
ELF3 | |
E. coli-derived recombinant human ELF3 Met1-Gly173 Accession # P78245 | |
25 μg | |
Primary | |
Detects human ELF3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human ELF5 is observed. | |
Human | |
Purified |
Immunohistochemistry | |
662516 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
E74-like factor 3, E74-like factor 3 (ets domain transcription factor, epithelial-specific ), Epithelial-restricted with serine box, epithelial-specific), Epithelium-restricted Ets protein ESX, Epithelium-specific Ets transcription factor 1, EPR1, EPR-1, ERT, ESX, ets domain transcription factor, serine box (epithelial-specific), ETS-related transcription factor Elf-3, JEN, serine box | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1999 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction