Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EMMPRIN/CD147 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB9721100
This item is not returnable.
View return policy
Description
EMMPRIN/CD147 Monoclonal antibody specifically detects EMMPRIN/CD147 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, KnockDownSpecifications
EMMPRIN/CD147 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
5F7, basigin, basigin (Ok blood group), BSG, CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor | |
Human embryonic kidney cell HEK293-derived human EMMPRIN/CD147 protein, Glu138-Ala323, Accession # P35613.2 | |
100 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2b |
Western Blot, Immunohistochemistry, Immunocytochemistry, KnockDown | |
1036020 | |
Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL, Knockout Validated 2 ug/mL | |
P35613.2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
682 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction