Learn More
Description
Specifications
Specifications
| Quantity | 1 Kit (for 15 Plates) |
| Species | Human |
| Conjugate | Biotin |
| Assay Range | 62.50 to 4000 pg/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 682 |
| Synonym | 5F7, basigin, basigin (Ok blood group), BSG, CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
