Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EMR2 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB4894G100UG
This item is not returnable.
View return policy
Description
EMR2 Monoclonal specifically detects EMR2 in Human samples. It is validated for Flow Cytometry.Specifications
EMR2 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD312, DKFZp781B135, egf-like module containing, mucin-like, hormone receptor-like 2, egf-like module containing, mucin-like, hormone receptor-like sequence 2, EGF-like module receptor 2, EGF-like module-containing mucin-like hormone receptor-like 2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human EMR2. Stains human EMR2-transfected cells but not the parental cell line. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2b |
Flow Cytometry | |
494025 | |
Flow Cytometry 0.25-1 μg/106 cells | |
AAI27006 | |
EMR2 | |
NS0 mouse myeloma cell line transfected with human EMR2, Gln24-Asn823, Accession # AAI27006 | |
100 μg | |
30817 | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction