Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EMR2 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB4894N100UG
This item is not returnable.
View return policy
Description
EMR2 Monoclonal specifically detects EMR2 in Human samples. It is validated for Flow Cytometry.Specifications
EMR2 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
AAI27006 | |
EMR2 | |
NS0 mouse myeloma cell line transfected with human EMR2 Gln24-Asn823 Accession # AAI27006 | |
RUO | |
30817 | |
Human | |
IgG2b |
Flow Cytometry | |
494025 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD312, DKFZp781B135, egf-like module containing, mucin-like, hormone receptor-like 2, egf-like module containing, mucin-like, hormone receptor-like sequence 2, EGF-like module receptor 2, EGF-like module-containing mucin-like hormone receptor-like 2 | |
Mouse | |
100 μg | |
Primary | |
Detects human EMR2 in EMR2-transfected cells but not in the parental cell line. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction