Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human EMR3 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB8496A025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8496A025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8496A025 Supplier R&D Systems Supplier No. FAB8496A025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

EMR3 Monoclonal specifically detects EMR3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen EMR3
Applications Flow Cytometry
Classification Monoclonal
Clone 908235
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. NP_115960
Gene Alias egf-like module containing, mucin-like, hormone receptor-like 3, EGF-like module receptor 3, EGF-like module-containing mucin-like hormone receptor-like 3, egf-like module-containing mucin-like receptor 3
Gene Symbols EMR3
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human EMR3 Met1-Tyr652 Accession # NP_115960
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84658
Test Specificity Stains human EMR3 transfected but not irrelevant transfected cells in flow cytometry.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.