Learn More
Invitrogen™ Human ENC-2 (aa 175-288) Control Fragment Recombinant Protein

Description
Highest antigen sequence indentity to the following orthologs: Mouse (87%), Rat (87%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82832 (PA5-82832. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Specifications
Specifications
Accession Number | Q9H0H3 |
Concentration | ≥5.0 mg/mL |
For Use With (Application) | Blocking Assay, Control |
Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
Gene ID (Entrez) | 64410 |
Name | Human ENC-2 (aa 175-288) Control Fragment |
Quantity | 100 μL |
Regulatory Status | RUO |
Gene Alias | 2810402K13Rik; BC027373; BTB/POZ KELCH domain protein; ectodermal-neural cortex 2; ectoderm-neural cortex protein 2; ectoderm-neural cortex-1; Enc-1; ENC2; ENC-2; FLJ12587; FLJ45015; kelch like family member 25; kelch-like 25; kelch-like family member 25; kelch-like protein 25; KLHL25; RGD1310815 |
Common Name | ENC-2 |
Show More |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.