Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Endocan/ESM-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1810
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1810 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1810 Supplier R&D Systems Supplier No. AF1810
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 3 publications

Endocan/ESM-1 Polyclonal specifically detects Endocan/ESM-1 in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen Endocan/ESM-1
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 1-4 ug/mL
Gene Accession No. Q9NQ30
Gene Alias endocan, endothelial cell-specific molecule 1, ESM1, ESM-1, IGFBP-rp6
Gene Symbols ESM1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Endocan/ESM-1 Trp20-Arg184 Accession # Q9NQ30
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11082
Test Specificity Detects human Endocan in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant mouse Endocan is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.