Learn More
Invitrogen™ Human Endogenous Retrovirus Polyclonal Antibody

Description
PA1-4109 detects human endogenous retrovirus in human samples. PA1-4109 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~55 kDa protein representing HERV-W in human placenta lysate. The PA1-4109 immunogen is a synthetic peptide corresponding to residues Q(42) R P G N I D A P(50) C of the HERV protein.
Specifications
Specifications
| Antigen | Human Endogenous Retrovirus |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene Alias | HERV |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to residues Q(42) R P G N I D A P(50) C of the HERV protein. |
| Purification Method | Antigen Affinity Chromatography |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.