Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Endogenous Retrovirus Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA123360
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA123360 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA123360 Supplier Invitrogen™ Supplier No. PA123360
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-23360 detects Endogenous Retrovirus in Virus samples.

Reacts with a replication-competent human Endogenous Retrovirus (HERV) isolated from placenta. HERVs are a class of multiple-sclerosis-associated retroviruses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human Endogenous Retrovirus
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias HERV
Host Species Rabbit
Immunogen Human Endogenous Retrovirus (HERV) isolated from placenta.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.