Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Endoglin/CD105 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB10971V100UG
This item is not returnable.
View return policy
Description
Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Human samples. It is validated for Flow Cytometry.Specifications
Endoglin/CD105 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q5T9B9 | |
ENG | |
Mouse myeloma cell line NS0-derived recombinant human Endoglin/CD105 Glu26-Gly586 Accession # Q5T9B9 | |
RUO | |
2022 | |
Human | |
IgG1 |
Flow Cytometry | |
166707 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, ENG, HHT1FLJ41744, ORW, ORW1 | |
Mouse | |
100 μg | |
Primary | |
Detects human Endoglin/CD105. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction