Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Endoglin/CD105 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$200.50 - $1885.00

Specifications

Antigen Endoglin/CD105
Clone 166713
Concentration LYOPH
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications ELISA
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB10972SP
GREENER_CHOICE
View Documents
R&D Systems
MAB10972SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB10972100
GREENER_CHOICE
View Documents
R&D Systems
MAB10972100
100 μg
Each for $514.50
Only null left
Add to Cart
 
MAB10972500
GREENER_CHOICE
View Documents
R&D Systems
MAB10972500
500 μg
Each for $1,885.00
Only null left
Add to Cart
 
Description

Description

Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

Endoglin/CD105
LYOPH
ELISA
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, ENG, HHT1FLJ41744, ORW, ORW1
ENG
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
166713
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Mouse
Human
P17813
2022
Mouse myeloma cell line NS0-derived recombinant human Endoglin/CD105 Glu26-Gly586 Accession # P17813
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Endoglin/CD105 in ELISAs. In sandwich ELISAs, no cross-reactivity was observed with recombinant mouse Endoglin, recombinant human (rh) Activin A, or rhTGF-beta 1.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.