Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Endoglin/CD105 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1097
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1097 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1097 Supplier R&D Systems Supplier No. BAF1097
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Endoglin/CD105 Polyclonal specifically detects Endoglin/CD105 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, ELISA Detection (Matched Antibody Pair).

Specifications

Antigen Endoglin/CD105
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. Q5T9B9
Gene Alias CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, ENG, HHT1FLJ41744, ORW, ORW1
Gene Symbols ENG
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Endoglin Glu26-Gly256 Accession # Q5T9B9
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2022
Test Specificity Detects human Endoglin in ELISAs and Western blots. In sandwich ELISAs, less than 0.2% cross-reactivity with recombinant mouse Endoglin, recombinant human (rh) Activin A, and rhTGF-beta 1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.