Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Endostatin Recombinant Protein, PeproTech®

Catalog No. p-7245754
Encompass_Preferred
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
500 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human Endostatin is a 20.2 kDa protein consisting of 184 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Endostatin is able to avoke non-uniform response for proliferation, cell mount and migration of entothelial cells, which different endostatin binding characteristic, leads to the assumption that endostatin effect is strongly dependent from endothelial cell type. Furthermore endostatin inhibits angiogenesis and tumor growth in vivo by inducing apoptosis in endothelial cells. The local delivery of endostatin seems to specifically affect tumor-associated microvessels by reduction of the vessel density, diameter and functionality. Tumor cell migration and invasion was greatly reduced in the endostatin treated animals. Endostatin is non-toxic and does not induce aquired drug resistance and has therefor become a potent new therapy strategy in solid neoplasias. This therapy appears to have a high potential not only for the treatment of gliomas, the most common brain tumors, but also in other tumors. The ability of endostatin to inhibit neoangiogenesis is mediated, at least in part, by Zn^+2 binding and elastase processing. Widespread endostatin expression was found in elactic fibers in vessel walls and in some other basement membrane zones. Endostatin is released by neurons to accumulate in amyloid plaques in Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P39060
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 80781
Molecular Weight (g/mol) 20.2 kDa
Name Human Endostatin
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias antiangiogenic agent; Col18a1; Collagen alpha-1(XVIII) chain; collagen type XVIII alpha 1 chain; collagen, type XVIII, alpha 1; endostatin; FLJ27325; FLJ34914; KNO; KNO1; KS; MGC74745; multi-functional protein MFP; NC1; Non-collagenous domain 1; procollagen, type XVIII, alpha 1
Common Name Endostatin
Gene Symbol COL18A1
Biological Activity Measured by its ability to inhibit HUVEC migration.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MHSHRDFQPV LHLVALNSPL SGGMRGIRGA DFQCFQQARA VGLAGTFRAF LSSRLQDLYS IVRRADRAAV PIVNLKDELL FPSWEALFSG SEGPLKPGAR IFSFDGKDVL RHPTWPQKSV WHGSDPNGRR LTESYCETWR TEAPSATGQA SSLLGGRLLG QSAASCHHAY IVLCIENSFM TASK
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.