Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human eNOS DuoSet ELISA

Catalog No. dy95005
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

This DuoSet ELISA Development kit contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant human Endothelial Nitric Oxide Synthase (eNOS).

The Human eNOS DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human eNOS.

Specifications

Assay Range 78.10 to 5,000pg/mL
Conjugate Biotin
Immunoassay Kit Format 96-well Strip Plate
Product Type Solid Phase Sandwich ELISA
For Use With (Application) ELISA
Gene ID (Entrez) 4846 (Human); 18127 (Mouse)
Kit Contents Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)
Quantity 1 Kit
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.