Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ENPP-1 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6136A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6136A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6136A Supplier R&D Systems Supplier No. FAB6136A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

ENPP-1 Polyclonal specifically detects ENPP-1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ENPP-1
Applications Flow Cytometry
Classification Polyclonal
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. Q13822 (ENPP-2) P22413 (ENPP-1)
Gene Alias ectonucleotide pyrophosphatase/phosphodiesterase 1, E-NPP 1, ENPP1, Ly-41 Antigen, M6S1, M6S1NPP1, Membrane component chromosome 6 surface marker 1, membrane component, chromosome 6, surface marker 1, NPP1, NPPS, NPPSalkaline phosphodiesterase 1, PC1, PC-1, PCA1, PCA1ARHR2, PDNP1, PDNP1ectonucleotide pyrophosphatase/phosphodiesterase family member 1, Phosphodiesterase I/nucleotide pyrophosphatase 1, plasma-cell membrane glycoprotein 1, Plasma-cell membrane glycoprotein PC-1
Gene Symbols ENPP1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human ENPP-2-1-2 Asp49-Trp144 (ENPP-2), Val191-Leu591 (ENPP-1), Asn532-Ile863 (ENPP-2). Cross-reactivity with human ENPP-2 was removed from the final product. Accession # Q13822 (ENPP-2) P22413 (ENPP-1)
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5167
Test Specificity Detects human ENPP-1 in direct ELISAs and Western blots. In direct ELISAs, less than 3% cross-reactivity with recombinant human ENPP-2 and recombinant mouse ENPP-2 is observed after removal of cross-reactivity with human ENPP-2.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.