Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Enteropeptidase/Enterokinase Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
| Antigen | Enteropeptidase/Enterokinase |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
Description
Enteropeptidase/Enterokinase Polyclonal specifically detects Enteropeptidase/Enterokinase in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Enteropeptidase/Enterokinase | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| EC 3.4.21, EC 3.4.21.9, Enterokinase, ENTK, ENTKenterokinase, MGC133046, protease, serine, 7 (enterokinase), PRSS7, PRSS7enteropeptidase, Serine protease 7, TMPRSS15, Transmembrane protease serine 15, transmembrane protease, serine 15 | |
| TMPRSS15 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| P98073 | |
| 5651 | |
| Mouse myeloma cell line NS0-derived recombinant human Enteropeptidase/Enterokinase Leu41-His1019 Accession # P98073 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Enteropeptidase/Enterokinase in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title