Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EpCAM/TROP-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 7 publications
$214.00 - $2020.00
Specifications
Antigen | EpCAM/TROP1 |
---|---|
Clone | 158210 |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Description
EpCAM/TROP1 Monoclonal specifically detects EpCAM/TROP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
EpCAM/TROP1 | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2, GA733-2EGP34, gp40, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A | |
EPCAM | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
158210 | |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P16422 | |
4072 | |
Mouse myeloma cell line NS0-derived recombinant human EpCAM/TROP-1, Gln24-Lys265, Accession # P16422 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human EpCAM/TROP-1 in direct ELISAs and Western blots. This antibody detects an epitope found in the extracellular domain between amino acids 136 and 265. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) ALCAM, rhBCAM, rhMCAM, rhNCAM-L1, or recombinant mouse (rm) OCAM is observed. In direct ELISAs, less than 5% cross-reactivity with rmEpCAM/TROP-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title